Learn More
Cayman Chemical 2'(3')-O-(5-Chloroanthraniloyl)adenosine-5'-O-triphosphate (sodium salt), Purity: ≥95% (mixture of isomers), 3.3 mg
SDP
Catalog No. 502918725
Encompass_Preferred
Fluorescent ATP derivatives; an inhibitor of mammalian AC1, AC2, and AC5 (Kis = 80, 490, and 45 nM, respectively), B. anthracis edema factor AC toxin (Ki = 170 nM), and B. pertussis CyaA (Ki = 350 nM)
Catalog No. 50-291-8725 Supplier Cayman Chemical Supplier No. 384493.3 MG
Fluorescent ATP derivatives; an inhibitor of mammalian AC1, AC2, and AC5 (Kis = 80, 490, and 45 nM, respectively), B. anthracis edema factor AC toxin (Ki = 170 nM), and B. pertussis CyaA (Ki = 350 nM)