Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone, 97+%

Catalog No. AAJ6644206
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6644206 5 g
AAJ66442ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6644206 Supplier Thermo Scientific Chemicals Supplier No. J6644206
Add to Cart

CAS: 30725-00-9 | C8H12O5 | 188.18 g/mol

2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone is used for the synthesis of C-nucleosides, prostanoids and GABA analogs via homochiral 2-piperidones. It is the starting material for the preparation of biochemical compounds including C-nucleosides, such as neplanocin A, and GABA analogs, via homochiral 2-piperidones.

Solubility
Soluble in methanol at 50mg/ml, acetone, dichloromethane, ethyl Acetate.

Notes
Soluble in methanol at 50mg/ml
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30725-00-9
Molecular Formula C8H12O5
Molecular Weight (g/mol) 188.18
MDL Number MFCD00080793
InChI Key NHHKFJCWLPPNCN-UHFFFAOYNA-N
Synonym 3ar,6r,6ar-6-hydroxymethyl-2,2-dimethyldihydrofuro 3,4-d 1,3 dioxol-4 3ah-one, 2,3-o-isopropylidene-d-ribonic gamma-lactone, 2,3-o-isopropylidene-d-ribono-1,4-lactone, 2,3-isopropylidene-d-ribonolactone, 3ar,6r,6ar-6-hydroxymethyl-2,2-dimethyl-dihydro-3ah-furo 3,4-d 1,3 dioxol-4-one, d-2,3-isopropylidene lyxono-1,4-lactone, pubchem10728, nhhkfjcwlppncn-hsuxutppsa, 2,3-o-isopropylidene-d-ribonic, a-lactone, 2,3-o-isopropylidene-d-ribonic-gama-lactone
PubChem CID 1268067
IUPAC Name 6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-one
SMILES CC1(C)OC2C(CO)OC(=O)C2O1

Specifications

Melting Point 135°C to 138°C
Color White
Quantity 5 g
Beilstein 10090
Solubility Information Soluble in methanol at 50mg/ml,acetone,dichloromethane,ethyl Acetate.
Optical Rotation −68° (c=2 in pyridine)
Formula Weight 188.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3-O-Isopropylidene-D-ribonic acid-1,4-lactone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.