Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-O-Isopropylidene-Guanosine, 98%, MP Biomedicals™

Catalog No. ICN10209901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10209901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10209901 Supplier MP Biomedicals, Inc Supplier No. 0210209901

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 362-76-5
Molecular Formula C13H17N5O5
Molecular Weight (g/mol) 323.31
MDL Number MFCD00057081
InChI Key XKPDAYWPKILAMO-IOSLPCCCSA-N
Synonym 2',3'-o-isopropylideneguanosine, 2-amino-9-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl-1h-purin-6 9h-one, 2',3'-o-1-methylethylidene guanosine, 2',3'-o-1-methylethylidene-guanosine, 9-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-2-amino-3h-purin-6-one, 9-3ar,4r,6r,6ar-6-hydroxymethyl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl-2-amino-1h-purin-6-one, 2', 3'-isopropylideneguanosine, 2'-o,3'-o-isopropylideneguanosine, 2',3'-o-1-methylethyliden-guanosin, 2/',3/'-o-1-methylethylidene guanosine
PubChem CID 6541409
IUPAC Name 9-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl]-2-amino-6,9-dihydro-3H-purin-6-one
SMILES CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)N1C=NC2=C1NC(N)=NC2=O

Specifications

Color White
Quantity 1 g
Physical Form Powder
Assay Percent Range 98%
Formula Weight 323.3
Chemical Name or Material 2', 3'-O-Isopropylidene-Guanosine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.