Learn More
Cayman Chemical 2'(3')-O-(N'-Methylanthraniloyl)guanosine-5'-O-triphosphate (sodium salt), Purity: ≥95% (mixture of isomers), 3.3 mg
SDP
Catalog No. 502917224
Encompass_Preferred
Fluorescent GTP derivatives; inhibits mammalian AC1, AC2, and AC5 (Kis = 90, 610, and 53 nM, respectively), B. pertussis AC toxin CyaA (Kis = 5.9 and 16 µM, in the presence of manganese and magnesium, respectively), and B. anthracis AC toxin edema factor (Kis = 2.49 and 4.7 µM in the presence of manganese and magnesium, respectively)
Catalog No. 50-291-7224 Supplier Cayman Chemical Supplier No. 384693.3 MG
Fluorescent GTP derivatives; inhibits mammalian AC1, AC2, and AC5 (Kis = 90, 610, and 53 nM, respectively), B. pertussis AC toxin CyaA (Kis = 5.9 and 16 µM, in the presence of manganese and magnesium, respectively), and B. anthracis AC toxin edema factor (Kis = 2.49 and 4.7 µM in the presence of manganese and magnesium, respectively)