Learn More
Cayman Chemical 2'(3')-O-Anthraniloylguanosine-5'-O-triphosphate (sodium salt), Purity: ≥95% (mixture of isomers), 3.2 mg
SDP
Catalog No. 502918375
Encompass_Preferred
Fluorescent GTP derivatives; inhibits mammalian AC1 (Ki = 10 nM in the presence of manganese), B. anthracis AC toxin edema factor (Kis = 4.1 and 13 µM in the presence of manganese and magnesium, respectively), and B. pertussis AC toxin CyaA (Ki = 29 µM in the presence of manganese)
Catalog No. 50-291-8375 Supplier Cayman Chemical Supplier No. 388123.2 MG
Fluorescent GTP derivatives; inhibits mammalian AC1 (Ki = 10 nM in the presence of manganese), B. anthracis AC toxin edema factor (Kis = 4.1 and 13 µM in the presence of manganese and magnesium, respectively), and B. pertussis AC toxin CyaA (Ki = 29 µM in the presence of manganese)