Learn More
Echelon Biosciences Research Labs 2,3-OXIDOSQUALENE 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs S030110MG
Encompass
2,3-Oxidosqualene is an important intermediate in the biosynthesis of membrane sterols, being converted to lanosterol by 2,3-oxidosqualene cyclase. This item is non returnable
Catalog No. 50-720-759