Learn More
Medchemexpress LLC 2-(3-oxo-1-phenyl-3-(4-(trifluoromethyl)phenyl)propyl)malonic acid | 1221962-86-2 | 98.0% | 10 MG
SDP
Catalog No. 5000230679
Encompass_Preferred
PS210 is a small-molecule activator of PDK1 provided for research use. It binds the PIF-binding pocket of PDK1 and is supplied as a solid or as a 10 mM solution in DMSO for in vitro studies.
  • Small-molecule PDK1 activator for research use.
  • Solid and 10 mM DMSO solution formats available.
  • High purity (~98.0%) confirmed by supplier analytical data.
  • Molecular weight 380.31 g/mol; formula C19H15F3O5.
  • Supplied in small milligram quantities suitable for biochemical assays.
Catalog No. 50-002-30679 Supplier Medchemexpress LLC Supplier No. HY12162910MG
PS210 is a small-molecule activator of PDK1 provided for research use. It binds the PIF-binding pocket of PDK1 and is supplied as a solid or as a 10 mM solution in DMSO for in vitro studies.
  • Small-molecule PDK1 activator for research use.
  • Solid and 10 mM DMSO solution formats available.
  • High purity (~98.0%) confirmed by supplier analytical data.
  • Molecular weight 380.31 g/mol; formula C19H15F3O5.
  • Supplied in small milligram quantities suitable for biochemical assays.