Learn More
Medchemexpress LLC 2-(3-oxo-1-phenyl-3-(4-(trifluoromethyl)phenyl)propyl)malonic acid | 1221962-86-2 | 98.0% | 100 MG
SDP
Catalog No. 5000234742
Encompass_Preferred
PS210 is a selective small-molecule activator of 3-phosphoinositide-dependent protein kinase-1 (PDK1), reported to bind the PIF-binding pocket with a Kd of approximately 3 μM. It is supplied as a high-purity research reagent available as solid packs and as a 10 mM solution in DMSO.
  • Selective activation of PDK1, reported Kd ≈ 3 μM.
  • High purity suitable for biochemical and cellular research.
  • Available as solids (1 mg-500 mg) and 10 mM solution in DMSO.
  • Stable as powder at -20°C for up to 3 years.
  • Molecular formula C19H15F3O5, molecular weight 380.31 g/mol.
Catalog No. 50-002-34742 Supplier Medchemexpress LLC Supplier No. HY121629100MG
PS210 is a selective small-molecule activator of 3-phosphoinositide-dependent protein kinase-1 (PDK1), reported to bind the PIF-binding pocket with a Kd of approximately 3 μM. It is supplied as a high-purity research reagent available as solid packs and as a 10 mM solution in DMSO.
  • Selective activation of PDK1, reported Kd ≈ 3 μM.
  • High purity suitable for biochemical and cellular research.
  • Available as solids (1 mg-500 mg) and 10 mM solution in DMSO.
  • Stable as powder at -20°C for up to 3 years.
  • Molecular formula C19H15F3O5, molecular weight 380.31 g/mol.