Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(3-oxo-1-phenyl-3-(4-(trifluoromethyl)phenyl)propyl)malonic acid | 1221962-86-2 | 98.3% | 25 MG
SDP

Catalog No. 5000232580
Encompass_Preferred

PS210 is a small-molecule activator of 3-phosphoinositide-dependent protein kinase-1 (PDK1) that engages the PIF-binding pocket and is used as a research reagent to study PDK1 signaling.

  • Potent and selective PDK1 activator for research use.
  • CAS number 1221962-86-2 for unambiguous identification.
  • Molecular formula C19H15F3O5 and molecular weight 380.31 g·mol⁻¹.
  • High purity (≈98%) suitable for biochemical assays.
  • Supplied as a solid powder; recommended storage at -20°C for long-term stability.
  • Available in small research pack sizes (25 mg) for laboratory studies.

Catalog No. 50-002-32580 Supplier Medchemexpress LLC Supplier No. HY12162925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PS210 is a small-molecule activator of 3-phosphoinositide-dependent protein kinase-1 (PDK1) that engages the PIF-binding pocket and is used as a research reagent to study PDK1 signaling.

  • Potent and selective PDK1 activator for research use.
  • CAS number 1221962-86-2 for unambiguous identification.
  • Molecular formula C19H15F3O5 and molecular weight 380.31 g·mol⁻¹.
  • High purity (≈98%) suitable for biochemical assays.
  • Supplied as a solid powder; recommended storage at -20°C for long-term stability.
  • Available in small research pack sizes (25 mg) for laboratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.