Learn More
Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | 99.1% | 100.12 | 100 MG
SDP
Catalog No. 5000452170
Encompass_Preferred
2,3-Pentanedione (Standard) is the analytical standard of 2,3-Pentanedione. It is intended for research and analytical applications. It is a common constituent of synthetic flavorings and is used to impart a butter, strawberry, caramel, fruit, rum, or cheese flavor in beverages, ice cream, candy, baked goods, gelatins, and puddings. It also occurs naturally as a fermentation product in beer, wine, and yogurt and is released during roasting of coffee beans.
  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments in hplc, gc, and ms.
Catalog No. 50-004-52170 Supplier Medchemexpress LLC Supplier No. HYW012998R100MG
2,3-Pentanedione (Standard) is the analytical standard of 2,3-Pentanedione. It is intended for research and analytical applications. It is a common constituent of synthetic flavorings and is used to impart a butter, strawberry, caramel, fruit, rum, or cheese flavor in beverages, ice cream, candy, baked goods, gelatins, and puddings. It also occurs naturally as a fermentation product in beer, wine, and yogurt and is released during roasting of coffee beans.
  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments in hplc, gc, and ms.