Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | 99.7% | 100.12 | 1 KG
SDP

Catalog No. 5000451994
Encompass_Preferred

2,3-Pentanedione is a light yellow to green yellow liquid with a molecular formula of C5H8O2 and a molecular weight of 100.12. It has a purity of 99.74% as determined by GC. This product is intended for research use only.

  • Light yellow to green yellow liquid
  • High purity (99.74%)
  • Molecular formula: C5H8O2
  • Molecular weight: 100.12
  • Long shelf life in pure form (3 years at -20°C, 2 years at 4°C)
  • Suitable for storage in solvent

Catalog No. 50-004-51994 Supplier Medchemexpress LLC Supplier No. HYW0129981KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-Pentanedione is a light yellow to green yellow liquid with a molecular formula of C5H8O2 and a molecular weight of 100.12. It has a purity of 99.74% as determined by GC. This product is intended for research use only.

  • Light yellow to green yellow liquid
  • High purity (99.74%)
  • Molecular formula: C5H8O2
  • Molecular weight: 100.12
  • Long shelf life in pure form (3 years at -20°C, 2 years at 4°C)
  • Suitable for storage in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.