Learn More
Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | 99.7% | 100.12 | 1 ML
SDP
Catalog No. 5000451391
Encompass_Preferred
2,3-Pentanedione is a chemical compound often found as a synthetic flavoring agent, contributing to butter, strawberry, caramel, fruit, rum, or cheese flavors in various food products. It naturally occurs during fermentation in products like beer and yogurt, and is released when coffee beans are roasted. Intended for laboratory research and manufacturing, this product requires handling by experienced personnel due to its properties.
  • Exhibits cytotoxicity in N2a cells and inhibits cell viability.
  • Induces severe inflammatory responses, including necrotizing rhinitis, tracheitis, and bronchitis, in Sprague-Dawley rats.
  • Causes olfactory neuroepithelial injury and changes in brain gene expression in rats.
  • Classified as a flammable liquid and vapor.
  • May cause an allergic skin reaction.
  • May cause organ damage through prolonged or repeated exposure.
  • Harmful to aquatic life.
Catalog No. 50-004-51391 Supplier Medchemexpress LLC Supplier No. HYW01299810MM/1ML
2,3-Pentanedione is a chemical compound often found as a synthetic flavoring agent, contributing to butter, strawberry, caramel, fruit, rum, or cheese flavors in various food products. It naturally occurs during fermentation in products like beer and yogurt, and is released when coffee beans are roasted. Intended for laboratory research and manufacturing, this product requires handling by experienced personnel due to its properties.
  • Exhibits cytotoxicity in N2a cells and inhibits cell viability.
  • Induces severe inflammatory responses, including necrotizing rhinitis, tracheitis, and bronchitis, in Sprague-Dawley rats.
  • Causes olfactory neuroepithelial injury and changes in brain gene expression in rats.
  • Classified as a flammable liquid and vapor.
  • May cause an allergic skin reaction.
  • May cause organ damage through prolonged or repeated exposure.
  • Harmful to aquatic life.