Learn More
Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | 99.7% | 100.12 | 100 G
SDP
Catalog No. 5000454138
Encompass_Preferred
2,3-Pentanedione is a common constituent of synthetic flavorings used to impart butter, strawberry, caramel, fruit, rum, or cheese flavor in various foods and beverages. It also occurs naturally as a fermentation product in beer, wine, and yogurt, and is released during coffee bean roasting. This product is for research use only.
  • Used to impart butter, strawberry, caramel, fruit, rum, or cheese flavor.
  • Naturally occurs as a fermentation product in beer, wine, and yogurt.
  • Released during coffee bean roasting.
  • Exhibits cytotoxicity in N2a cells in vitro.
  • Causes inflammatory responses in rats in vivo.
Catalog No. 50-004-54138 Supplier Medchemexpress LLC Supplier No. HYW012998100G
2,3-Pentanedione is a common constituent of synthetic flavorings used to impart butter, strawberry, caramel, fruit, rum, or cheese flavor in various foods and beverages. It also occurs naturally as a fermentation product in beer, wine, and yogurt, and is released during coffee bean roasting. This product is for research use only.
  • Used to impart butter, strawberry, caramel, fruit, rum, or cheese flavor.
  • Naturally occurs as a fermentation product in beer, wine, and yogurt.
  • Released during coffee bean roasting.
  • Exhibits cytotoxicity in N2a cells in vitro.
  • Causes inflammatory responses in rats in vivo.