Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | 99.74% | 100.12 | 25MG
SDP

Catalog No. 5000366650
Encompass_Preferred

2,3-Pentanedione is a common constituent found in synthetic flavorings, used to impart butter, strawberry, caramel, fruit, rum, or cheese flavors in various food and beverage products such as ice cream, candy, baked goods, gelatins, and puddings. It also occurs naturally as a fermentation product in beer, wine, and yogurt, and is released during the roasting of coffee beans.

  • Purity: 99.74%
  • Appearance: light yellow to green yellow liquid (density: 0.9571 g/cm³)
  • Occurs naturally as a fermentation product and is also synthesized for flavoring applications
  • Versatile flavoring agent that imparts a variety of desirable flavors including butter, strawberry, caramel, fruit, rum, and cheese
  • Suitable for use in beverages, ice cream, candy, baked goods, gelatins, and puddings

Catalog No. 50-003-66650 Supplier Medchemexpress LLC Supplier No. 25MGHYW012998R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-Pentanedione is a common constituent found in synthetic flavorings, used to impart butter, strawberry, caramel, fruit, rum, or cheese flavors in various food and beverage products such as ice cream, candy, baked goods, gelatins, and puddings. It also occurs naturally as a fermentation product in beer, wine, and yogurt, and is released during the roasting of coffee beans.

  • Purity: 99.74%
  • Appearance: light yellow to green yellow liquid (density: 0.9571 g/cm³)
  • Occurs naturally as a fermentation product and is also synthesized for flavoring applications
  • Versatile flavoring agent that imparts a variety of desirable flavors including butter, strawberry, caramel, fruit, rum, and cheese
  • Suitable for use in beverages, ice cream, candy, baked goods, gelatins, and puddings

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.