Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-Pentanedione | 600-14-6 | HY-W012998 | 99.7% | 100.12 | 50 G
SDP

Catalog No. 5000451995
Encompass_Preferred

2,3-Pentanedione is a common constituent of synthetic flavorings used to impart butter, strawberry, caramel, fruit, rum, or cheese flavors in various foods and beverages such as ice cream, candy, baked goods, gelatins, and puddings. It also occurs naturally as a fermentation product in beer, wine, and yogurt and is released during the roasting of coffee beans. For research use only.

  • Appearance: Liquid (density: 0.9571 g/cm³).
  • Color: Light yellow to green yellow.
  • Structure classification: Ketones, Aldehydes, Acids.
  • Initial source: Plants other families.
  • Shipping: Room temperature in continental US.
  • Storage (pure form): -20°C for 3 years or 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-004-51995 Supplier Medchemexpress LLC Supplier No. HYW01299850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-Pentanedione is a common constituent of synthetic flavorings used to impart butter, strawberry, caramel, fruit, rum, or cheese flavors in various foods and beverages such as ice cream, candy, baked goods, gelatins, and puddings. It also occurs naturally as a fermentation product in beer, wine, and yogurt and is released during the roasting of coffee beans. For research use only.

  • Appearance: Liquid (density: 0.9571 g/cm³).
  • Color: Light yellow to green yellow.
  • Structure classification: Ketones, Aldehydes, Acids.
  • Initial source: Plants other families.
  • Shipping: Room temperature in continental US.
  • Storage (pure form): -20°C for 3 years or 4°C for 2 years.
  • Storage (in solvent): -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.