Learn More
Medchemexpress LLC 2,3-Pentanedione (Standard) | 600-14-6 | 99.1% | 100.12 | 500 MG
SDP
Catalog No. 5000451193
Encompass_Preferred
2,3-Pentanedione (Standard) is an analytical standard intended for research and analytical applications. It is recognized for its role as a flavoring agent in various foods and beverages, contributing flavors such as butter, strawberry, caramel, fruit, rum, or cheese. The compound is naturally found as a fermentation product in beer, wine, and yogurt, and is also released during the roasting of coffee beans. It is typically used in qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Common constituent of synthetic flavorings
  • Used to impart butter, strawberry, caramel, fruit, rum, or cheese flavor
  • Occurs naturally as a fermentation product in beer, wine, and yogurt
  • Released during the roasting of coffee beans
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-004-51193 Supplier Medchemexpress LLC Supplier No. HYW012998R500MG
2,3-Pentanedione (Standard) is an analytical standard intended for research and analytical applications. It is recognized for its role as a flavoring agent in various foods and beverages, contributing flavors such as butter, strawberry, caramel, fruit, rum, or cheese. The compound is naturally found as a fermentation product in beer, wine, and yogurt, and is also released during the roasting of coffee beans. It is typically used in qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Common constituent of synthetic flavorings
  • Used to impart butter, strawberry, caramel, fruit, rum, or cheese flavor
  • Occurs naturally as a fermentation product in beer, wine, and yogurt
  • Released during the roasting of coffee beans
  • Used in HPLC, GC, and MS research experiments