Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Pentanedione 97.0+%, TCI America™
SDP

Catalog No. P0051500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0051500ML 500 mL
P005125ML 25 mL
P0051100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0051500ML Supplier TCI America Supplier No. P0051500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 600-14-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00009313
InChI Key TZMFJUDUGYTVRY-UHFFFAOYSA-N
Synonym 2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
PubChem CID 11747
ChEBI CHEBI:52774
IUPAC Name pentane-2,3-dione
SMILES CCC(=O)C(C)=O

Specifications

Melting Point -52°C
Color Yellow
Boiling Point 108°C
Quantity 500 mL
UN Number 1224
Formula Weight 100.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Pentanedione
EINECSNumber (2)-0563&(2)-2474
RTECSNumber SA1850000
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.