Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-piperidinedicarboxylic acid, (2R,3S)-rel- | 46026-75-9 | 98.0% | 25 MG
SDP

Catalog No. 5000233713
Encompass_Preferred

cis-Piperidine-2,3-dicarboxylic acid is a high-purity research reagent described as an NMDA receptor agonist, supplied as a small molecule for laboratory pharmacology and neurobiology studies.

  • High purity: 98.0%.
  • Suitable for in vitro receptor pharmacology and screening assays.
  • Available in small milligram quantities for screening and pilot studies.
  • Molecular weight 173.17 g/mol.
  • Molecular formula C7H11NO4.
  • Provided with a certificate of analysis for batch-specific data.

Catalog No. 50-002-33713 Supplier Medchemexpress LLC Supplier No. HY12196425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

cis-Piperidine-2,3-dicarboxylic acid is a high-purity research reagent described as an NMDA receptor agonist, supplied as a small molecule for laboratory pharmacology and neurobiology studies.

  • High purity: 98.0%.
  • Suitable for in vitro receptor pharmacology and screening assays.
  • Available in small milligram quantities for screening and pilot studies.
  • Molecular weight 173.17 g/mol.
  • Molecular formula C7H11NO4.
  • Provided with a certificate of analysis for batch-specific data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.