Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)ethanamine trifluoroacetate salt

Catalog No. CBR010751G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBR010751G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBR010751G Supplier Sigma Aldrich Supplier No. CBR010751G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)ethanamine trifluoroacetate salt

Specifications

Molecular Formula C11H11F3N4O3
Linear Formula C11H11F3N4O3
Quantity 250 mg
MDL Number MFCD12028447
Molecular Weight (g/mol) 304.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.