Learn More
Medchemexpress LLC 2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid | 39091-01-5 | 96.8% | 220.25 | 500 MG
SDP
Catalog No. 5000453696
Encompass_Preferred
2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid is a synthetic intermediate primarily utilized in pharmaceutical synthesis. This compound serves as a crucial building block in the development of more complex chemical structures for drug discovery and other related applications, facilitating the creation of novel compounds with desired biological activities. Its specific structural properties make it valuable in various synthetic pathways.
  • Synthetic intermediate for pharmaceutical synthesis.
  • Useful in drug development.
  • Serves as a building block for complex molecules.
  • Valuable in various synthetic pathways.
Catalog No. 50-004-53696 Supplier Medchemexpress LLC Supplier No. HYW012428500MG
2-(3-Pyridyl)-4-methyl-thiazole-5-carboxylic acid is a synthetic intermediate primarily utilized in pharmaceutical synthesis. This compound serves as a crucial building block in the development of more complex chemical structures for drug discovery and other related applications, facilitating the creation of novel compounds with desired biological activities. Its specific structural properties make it valuable in various synthetic pathways.
  • Synthetic intermediate for pharmaceutical synthesis.
  • Useful in drug development.
  • Serves as a building block for complex molecules.
  • Valuable in various synthetic pathways.