Learn More
Medchemexpress LLC HY-112535 5mg Medchemexpress, α-2,3-sialyltransferase-IN-1 CAS:881179-06-2 Purity:>98%
SDP
Catalog No. 501969101
Encompass_Preferred
Medchemexpress, HY-112535 5mg α-2,3-sialyltransferase-IN-1 CAS:881179-06-2 α-2,3-sialyltransferase-IN-1 (Lith-O-Asp analog) is a noncompetitive α-2,3-sialyltransferase inhibitor with an IC50 of 6 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9101 Supplier Medchemexpress LLC Supplier No. HY1125355MG
Medchemexpress, HY-112535 5mg α-2,3-sialyltransferase-IN-1 CAS:881179-06-2 α-2,3-sialyltransferase-IN-1 (Lith-O-Asp analog) is a noncompetitive α-2,3-sialyltransferase inhibitor with an IC50 of 6 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.