Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(3-(Trifluoromethyl)benzamido)acetic acid | 17794-48-8 | 99.3% | C10H8F3NO3 | 500 MG
SDP

Catalog No. 5000434355
Encompass_Preferred

2-(3-(Trifluoromethyl)benzamido)acetic acid is a drug intermediate for the synthesis of various active compounds. This product, with a purity of 99.3%, appears as a white to light yellow solid and is stable under recommended storage conditions.

  • Purity (LCMS): 99.3%
  • Appearance: White to light yellow solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Shipping at room temperature if less than 2 weeks

Catalog No. 50-004-34355 Supplier Medchemexpress LLC Supplier No. HY75204500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(3-(Trifluoromethyl)benzamido)acetic acid is a drug intermediate for the synthesis of various active compounds. This product, with a purity of 99.3%, appears as a white to light yellow solid and is stable under recommended storage conditions.

  • Purity (LCMS): 99.3%
  • Appearance: White to light yellow solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month
  • Keep container tightly sealed in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
  • Shipping at room temperature if less than 2 weeks

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.