Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-[3-(trifluoromethyl)phenoxy]nicotinic acid

Catalog No. CDS015113 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS015113 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS015113 Supplier Sigma Aldrich Supplier No. CDS015113100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-chloro-4-iodobutane is a halogenated hydrocarbon. it is an α,-d-dihaloalkane and undergoes electrogenerated nickel(i) salen (n,nα-bis(salicylidene)ethylenediamine) catalyzed reduction to afford 1,8-dichlorooctane. electrochemical reduction of 1-chloro-4-iodobutane at glassy carbon cathode has been investigated by cyclic voltammetry and controlled-potential electrolysis.1-chloro-4-iodobutane may be used in the following studies:
  • Preparation of 6-hendecenoic acid
  • Catalytic asymmetric synthesis of levobupivacaine
  • Synthesis of alkaloids such as deoxyvasicinone, mackinazolinone

Specifications

CAS 36701-89-0
Molecular Formula C13H8F3NO3
Linear Formula C13H8F3NO3
MDL Number MFCD01938784
Quantity 5 mL
Molecular Weight (g/mol) 283.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.