Learn More
Electronic Imaging Materials Inc 2.36 Inch x 300 Meter T84 Premium Resin Ribbon - EIMINC
SDP
Catalog No. NC1399574
Encompass_Preferred
T84—2.36 inch x 300 meter anti-static, halogen-free ribbon is ultra-durable. Creates an excellent smudge, scratch, and solvent resistant image. High heat resistance—great for long-term labeling requirements. T84 resin ribbon eliminates the need for over-laminating in most situations. *Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra printer models ZT220/230, ZT410/420, ZD420, as well as TSC TTP247/345 and TSC TX series printers.
Catalog No. NC1399574 Supplier Electronic Imaging Materials Inc Supplier No. T84236300
T84—2.36 inch x 300 meter anti-static, halogen-free ribbon is ultra-durable. Creates an excellent smudge, scratch, and solvent resistant image. High heat resistance—great for long-term labeling requirements. T84 resin ribbon eliminates the need for over-laminating in most situations. *Note: make sure to find out if ribbon is compatible with your printer & label material before purchasing. Works in Zebra printer models ZT220/230, ZT410/420, ZD420, as well as TSC TTP247/345 and TSC TX series printers.