Learn More
Cayman Chemical 2,3-bis (4-Hydroxyphenyl) Propionitrile, Purity: ≥98%, CAS: 1428-67-7, 100 mg
SDP
Catalog No. 502887369
Encompass_Preferred
An ERβ Selective agonist; exhibits 70-fold higher relative binding affinity for ERβ versus ERα
Catalog No. 50-288-7369 Supplier Cayman Chemical Supplier No. 10008842100 MG
An ERβ Selective agonist; exhibits 70-fold higher relative binding affinity for ERβ versus ERα