Learn More
Cayman Chemical 2 3bis 4Hydroxyphenyl Propio
SDP
Supplier:  Cayman Chemical 100088425 MG
Encompass_Preferred
An ERβ Selective agonist; exhibits 70-fold higher relative binding affinity for ERβ versus ERα
Catalog No. 50-289-0033