Learn More
Cayman Chemical 2 3cyclIc NADP sodIum sa 7.3mg
SDP
Catalog No. 502919141
Encompass_Preferred
A substrate for CNP; has been used in coupled enzyme assays to quantify CNP activity; increases calcium overload-induced calcium release and prevents calcium-induced swelling in rat brain mitochondria at 5 µM
Catalog No. 50-291-9141 Supplier Cayman Chemical Supplier No. 383887.3 MG
A substrate for CNP; has been used in coupled enzyme assays to quantify CNP activity; increases calcium overload-induced calcium release and prevents calcium-induced swelling in rat brain mitochondria at 5 µM