Learn More
Cayman Chemical 2 3Dehydro3 4dIhydro IvRm 10mg
SDP
Catalog No. 502993683
Encompass_Preferred
An anthelmintic and ivermectin analog; has activity against L. amazonensis promastigotes and amastigotes (IC50s = 13.8 and 3.6 µM, respectively) without inducing cytotoxicity to macrophages (IC50 = 65.5 µM)
Catalog No. 50-299-3683 Supplier Cayman Chemical Supplier No. 2384910 MG
An anthelmintic and ivermectin analog; has activity against L. amazonensis promastigotes and amastigotes (IC50s = 13.8 and 3.6 µM, respectively) without inducing cytotoxicity to macrophages (IC50 = 65.5 µM)