Learn More
Cayman Chemical 2 3DICLOrophenylpIpRazIn 10mg
SDP
Catalog No. 502955439
Encompass_Preferred
An analytical reference standard categorized as a piperazine; precursor in the synthesis of aripiprazole; has been found as an impurity in commercial formulations of aripiprazole; intended for research and forensic applications
Catalog No. 50-295-5439 Supplier Cayman Chemical Supplier No. 1429610 MG
An analytical reference standard categorized as a piperazine; precursor in the synthesis of aripiprazole; has been found as an impurity in commercial formulations of aripiprazole; intended for research and forensic applications