Learn More
Cayman Chemical 2,3-Dimethylmethcathinone (hydrochloride), Purity: ≥95%, CAS: 1797981-99-7, 50 mg
SDP
Catalog No. 502951346
Encompass_Preferred
A potential designer drug that is structurally related to 4-MMC; intended for research and forensic applications
Catalog No. 50-295-1346 Supplier Cayman Chemical Supplier No. 1122550 MG
A potential designer drug that is structurally related to 4-MMC; intended for research and forensic applications