Learn More
Enzo Life Sciences 2,3-Dinor-6-keto-prostaglandin F1alpha-20,20,20-d3, (50µg)
SDP
Catalog No. NC9931702
Encompass_Preferred
Ultra-pure. Alternative Name: 2,3-Dinor-6-keto-PGF1alpha-d3. Formula: C18H27D3O6. MW: 345.5. Purity: ≥98% (TLC). Appearance: Liquid. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Long Term Storage: -20°C
Catalog No. NC9931702 Supplier Enzo Life Sciences Supplier No. BMLPG0400050
Ultra-pure. Alternative Name: 2,3-Dinor-6-keto-PGF1alpha-d3. Formula: C18H27D3O6. MW: 345.5. Purity: ≥98% (TLC). Appearance: Liquid. Use/Stability: Store, as supplied, at -20°C for up to 1 year. Long Term Storage: -20°C