Learn More
Medchemexpress LLC 2(3h)-benzothiazolone, 6-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-3-methyl | 142477-34-7 | 97.1% | 383.51 g/mol | C21H25N3O2S | 1 MG
SDP
Catalog No. 5000214116
Encompass_Preferred
A selective small-molecule research ligand for the 5-HT1A serotonin receptor that also exhibits activity at adrenergic α1 and dopamine D2 receptors. Supplied as a white to off-white solid with supporting analytical documentation (COA and datasheet) for preclinical pharmacology and receptor profiling studies.
  • High affinity for 5-HT1A receptor, IC50 ≈ 2 nM.
  • Measured off-target activity at adrenergic α1 and dopamine D2 receptors (IC50 ≈ 10 nM and 40 nM).
  • Supplied as a stable solid suitable for frozen or refrigerated storage.
  • Certificate of analysis and purity data available to support assay reproducibility.
  • Useful as a pharmacological tool for serotonin receptor profiling and CNS research.
Catalog No. 50-002-14116 Supplier Medchemexpress LLC Supplier No. HY1002901MG
A selective small-molecule research ligand for the 5-HT1A serotonin receptor that also exhibits activity at adrenergic α1 and dopamine D2 receptors. Supplied as a white to off-white solid with supporting analytical documentation (COA and datasheet) for preclinical pharmacology and receptor profiling studies.
  • High affinity for 5-HT1A receptor, IC50 ≈ 2 nM.
  • Measured off-target activity at adrenergic α1 and dopamine D2 receptors (IC50 ≈ 10 nM and 40 nM).
  • Supplied as a stable solid suitable for frozen or refrigerated storage.
  • Certificate of analysis and purity data available to support assay reproducibility.
  • Useful as a pharmacological tool for serotonin receptor profiling and CNS research.