Learn More
Chemglass Life Sciences Chemglass Baffle Assembly for 2-3 L Bioreactor (M6 thread)
SDP
Catalog No. 502733317
Encompass
Chemglass baffle assembly designed to increase mixing efficiency in 2-3 L bioreactors. It mounts near the reactor wall and uses an M6 thread for secure attachment, creating turbulence to improve mixing and mass transfer.
  • Increases mixing efficiency inside the bioreactor.
  • Mounts near the reactor wall for optimum mixing performance.
  • M6 thread for secure attachment to compatible headplates.
  • Compatible with 2-3 L bioreactor vessels.
  • Constructed from stainless steel; related components are typically 316L with an electropolished 20 Ra finish.
Catalog No. 50-273-3317 Supplier Chemglass Life Sciences Supplier No. CLS138104
Chemglass baffle assembly designed to increase mixing efficiency in 2-3 L bioreactors. It mounts near the reactor wall and uses an M6 thread for secure attachment, creating turbulence to improve mixing and mass transfer.
  • Increases mixing efficiency inside the bioreactor.
  • Mounts near the reactor wall for optimum mixing performance.
  • M6 thread for secure attachment to compatible headplates.
  • Compatible with 2-3 L bioreactor vessels.
  • Constructed from stainless steel; related components are typically 316L with an electropolished 20 Ra finish.