Learn More
BioSpec Products 2.3MM ZIRCONIA BEADS
SDP
Catalog No. NC2128806
Encompass_Preferred
Zirconia beads, our most dense ceramic bead, has a density twice that of regular glass beads. Sold in one pound bottles.Our Zirconia beads are great for tough tissues. Our ZX series have a density of 5.5 g/cc. Our ZXY series offer equal or better performance than our original ZX line but are Yttria stabilized and feature a higher density of 6.0 g/cc.Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value.
This item is not returnable. View return policy
Catalog No. NC2128806 Supplier BioSpec Products Supplier No. 11079123ZXY
Zirconia beads, our most dense ceramic bead, has a density twice that of regular glass beads. Sold in one pound bottles.Our Zirconia beads are great for tough tissues. Our ZX series have a density of 5.5 g/cc. Our ZXY series offer equal or better performance than our original ZX line but are Yttria stabilized and feature a higher density of 6.0 g/cc.Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value.