Learn More
Cayman Chemical 2'(3')-O-(6-Aminohexylcarbamoyl)guanosine-5'-O-triphosphate (sodium salt), Purity: ≥95% (mixture of isomers), 3.3 mg
SDP
Catalog No. 502918832
Encompass_Preferred
A mixture of GTP derivatives; has been used in the synthesis of europium (III) chelates for use in QRET assays in the study of GTPase nucleotide exchange kinetics
Catalog No. 50-291-8832 Supplier Cayman Chemical Supplier No. 386593.3 MG
A mixture of GTP derivatives; has been used in the synthesis of europium (III) chelates for use in QRET assays in the study of GTPase nucleotide exchange kinetics