Learn More
Cayman Chemical 2 3Pentylon Isomerhydroch 10mg
SDP
Catalog No. 502951521
Encompass_Preferred
A structural isomer of pentylone, having the methylenedioxy group attached at carbons 2 and 3 of the terminal phenyl group; intended for forensic and research applications
Catalog No. 50-295-1521 Supplier Cayman Chemical Supplier No. 1146310 MG
A structural isomer of pentylone, having the methylenedioxy group attached at carbons 2 and 3 of the terminal phenyl group; intended for forensic and research applications