Learn More
Cayman Chemical 2,3-Pentylone isomer (hydrochloride), Purity: ≥98%, CAS: 2749428-57-5, 50 mg
SDP
Catalog No. 502951303
Encompass_Preferred
A structural isomer of pentylone, having the methylenedioxy group attached at carbons 2 and 3 of the terminal phenyl group; intended for forensic and research applications
Catalog No. 50-295-1303 Supplier Cayman Chemical Supplier No. 1146350 MG
A structural isomer of pentylone, having the methylenedioxy group attached at carbons 2 and 3 of the terminal phenyl group; intended for forensic and research applications