Learn More
Medchemexpress LLC 2-[[(3R)-3-(4-fluorophenyl)-3-(4-phenylphenoxy)propyl]-methylamino]acetic acid hydrochloride | 200006-08-2 | MFCD09971056 | 98.2% | 429.91 | C24H25ClFNO3 | 1 MG
SDP
Catalog No. 5000239773
Encompass_Preferred
ALX-5407 hydrochloride is the hydrochloride salt of a selective glycine transporter 1 (GlyT1) inhibitor used in preclinical research to increase extracellular glycine and probe GlyT1 biology. It is provided as a high-purity research chemical for in vitro and in vivo studies.
  • Selective glycine transporter 1 inhibitor with high potency (IC50 ≈ 3 nM).
  • Hydrochloride salt form for improved solubility.
  • High purity (98.2%) suitable for research applications.
  • Available in small quantities for dose-response and pilot studies.
  • Molecular weight 429.91 and formula C24H25ClFNO3 for precise calculations.
Catalog No. 50-002-39773 Supplier Medchemexpress LLC Supplier No. HY10711A1MG
ALX-5407 hydrochloride is the hydrochloride salt of a selective glycine transporter 1 (GlyT1) inhibitor used in preclinical research to increase extracellular glycine and probe GlyT1 biology. It is provided as a high-purity research chemical for in vitro and in vivo studies.
  • Selective glycine transporter 1 inhibitor with high potency (IC50 ≈ 3 nM).
  • Hydrochloride salt form for improved solubility.
  • High purity (98.2%) suitable for research applications.
  • Available in small quantities for dose-response and pilot studies.
  • Molecular weight 429.91 and formula C24H25ClFNO3 for precise calculations.