Learn More
Medchemexpress LLC 2-(4-(1,2,4,5-tetrazin-3-yl)phenyl)acetic acid | 1380500-92-4 | MFCD24503098 | 98.7% | 216.20 g/mol | C10H8N4O2 | 25MG
SDP
Catalog No. 5000236571
Encompass_Preferred
Tetrazine-Ph-acid is a tetrazine-containing small-molecule linker used for bioorthogonal inverse electron-demand Diels-Alder (iEDDA) click chemistry and in PROTAC linker synthesis. The molecule is carboxylic-acid functionalized, enabling standard coupling reactions and further derivatization for medicinal chemistry and chemical biology research applications.
Catalog No. 50-002-36571 Supplier Medchemexpress LLC Supplier No. HY12448025MG
Tetrazine-Ph-acid is a tetrazine-containing small-molecule linker used for bioorthogonal inverse electron-demand Diels-Alder (iEDDA) click chemistry and in PROTAC linker synthesis. The molecule is carboxylic-acid functionalized, enabling standard coupling reactions and further derivatization for medicinal chemistry and chemical biology research applications.