Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid Pharmaceutical Impurity Standard, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014535 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4535 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4535 Supplier MilliporeSigma Supelco Supplier No. 6224850MG
Add to Cart
Add to Cart
Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 53949-53-4
Linear Formula C13H18O3
MDL Number MFCD08275586
Synonym 1-Hydroxyibuprofen; Ibuprofen impurity L (PhEur); 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 222.28
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.