Learn More
Medchemexpress LLC 2-(4-(1-(quinolin-6-ylmethyl)-1H-triazolo[4,5-b]pyrazin-6-yl)-1H-pyrazol-1-yl)ethanol | 956906-93-7 | ≥98.0% | 50 MG
SDP
Catalog No. 5000233428
Encompass_Preferred
PF-04217903 mesylate is the methanesulfonate salt of a selective ATP-competitive c-Met (MET) tyrosine kinase inhibitor used as a research reagent for preclinical studies. It is supplied as a solid salt form for biochemical and cell-based assays to evaluate c-Met signaling and inhibitor activity.
  • Selective ATP-competitive c-Met inhibitor for research applications.
  • Mesylate salt form to improve solubility and handling.
  • Typical purity ≥98.0% by HPLC.
  • Molecular weight 468.49 g/mol.
  • Intended for biochemical kinase assays and cell-based studies.
Catalog No. 50-002-33428 Supplier Medchemexpress LLC Supplier No. HY12017A50MG
PF-04217903 mesylate is the methanesulfonate salt of a selective ATP-competitive c-Met (MET) tyrosine kinase inhibitor used as a research reagent for preclinical studies. It is supplied as a solid salt form for biochemical and cell-based assays to evaluate c-Met signaling and inhibitor activity.
  • Selective ATP-competitive c-Met inhibitor for research applications.
  • Mesylate salt form to improve solubility and handling.
  • Typical purity ≥98.0% by HPLC.
  • Molecular weight 468.49 g/mol.
  • Intended for biochemical kinase assays and cell-based studies.