Learn More
Medchemexpress LLC 2,4(1H,3H)-Pyrimidinedione, 1-[[5-(2,4-difluoro-3-hydroxyphenyl)-1,3,4-thiadiazol-2-yl]methyl]-3-ethyl-5-methyl- | 2894848-07-6 | 99.8% | 380.37 | 100 MG
SDP
Catalog No. 5000361962
Encompass_Preferred
BI-3231 is a chemical probe that acts as a potent and selective inhibitor of hydroxysteroid 17β-dehydrogenase 13 (HSD17B13). With IC50 values of 1 nM for hHSD17B13 and 13 nM for mHSD17B13, it is being researched for its potential use in nonalcoholic steatohepatitis (NASH) and other liver diseases. This product is intended for research use only.
  • Potent and selective HSD17B13 inhibitor
  • Researched for nonalcoholic steatohepatitis (NASH) treatment
  • High metabolic stability in liver microsomes
  • Considerable hepatic exposure maintained over 48 hours in vivo
Catalog No. 50-003-61962 Supplier Medchemexpress LLC Supplier No. HY148814100MG
BI-3231 is a chemical probe that acts as a potent and selective inhibitor of hydroxysteroid 17β-dehydrogenase 13 (HSD17B13). With IC50 values of 1 nM for hHSD17B13 and 13 nM for mHSD17B13, it is being researched for its potential use in nonalcoholic steatohepatitis (NASH) and other liver diseases. This product is intended for research use only.
  • Potent and selective HSD17B13 inhibitor
  • Researched for nonalcoholic steatohepatitis (NASH) treatment
  • High metabolic stability in liver microsomes
  • Considerable hepatic exposure maintained over 48 hours in vivo