Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4(1H,3H)-pyrimidinedione, 1-β-D-arabinofuranosyl-5-methyl | 605-23-2 | MFCD00065474 | 98.0% | 258.23 g·mol⁻¹ | C10H14N2O6 | 5 MG
SDP

Catalog No. 5000223406
Encompass_Preferred

Thymine 1-β-D-arabinofuranoside is a thymine-derived nucleoside analogue used as a research reagent to study nucleoside metabolism and DNA synthesis inhibition. It is supplied as a characterized solid with high purity for in vitro biochemical and cell-based assays.

  • Purity 98.0%.
  • Molecular weight 258.23 g·mol⁻¹.
  • Chemical formula C10H14N2O6.
  • Supplied as a solid powder suitable for laboratory use.
  • Available in small milligram packaging for research applications.
  • Commonly used to investigate DNA synthesis and nucleoside metabolism.

Catalog No. 50-002-23406 Supplier Medchemexpress LLC Supplier No. HY1378485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymine 1-β-D-arabinofuranoside is a thymine-derived nucleoside analogue used as a research reagent to study nucleoside metabolism and DNA synthesis inhibition. It is supplied as a characterized solid with high purity for in vitro biochemical and cell-based assays.

  • Purity 98.0%.
  • Molecular weight 258.23 g·mol⁻¹.
  • Chemical formula C10H14N2O6.
  • Supplied as a solid powder suitable for laboratory use.
  • Available in small milligram packaging for research applications.
  • Commonly used to investigate DNA synthesis and nucleoside metabolism.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.