Learn More
Medchemexpress LLC 2,4(1H,3H)-pyrimidinedione-5,6-d2 | 24897-52-7 | 99.9% | 114.10 | C4H2D2N2O2 | 10 MG
SDP
Catalog No. 5000298987
Encompass_Preferred
Uracil-d2 is the deuterium-labeled analogue of uracil, used as an isotopic standard and tracer in analytical and metabolic studies. It is a white to off-white solid with molecular formula C4H2D2N2O2 and CAS number 24897-52-7.
  • Deuterium-labeled for mass spectrometry tracing and isotopic studies.
  • High reported purity (≈99.9%).
  • Suitable as an internal standard for quantitative assays.
  • Supplied in small research quantities appropriate for analytical applications.
  • Stable as a powder with recommended cold storage to preserve isotopic integrity.
Catalog No. 50-002-98987 Supplier Medchemexpress LLC Supplier No. HYI0960S410MG
Uracil-d2 is the deuterium-labeled analogue of uracil, used as an isotopic standard and tracer in analytical and metabolic studies. It is a white to off-white solid with molecular formula C4H2D2N2O2 and CAS number 24897-52-7.
  • Deuterium-labeled for mass spectrometry tracing and isotopic studies.
  • High reported purity (≈99.9%).
  • Suitable as an internal standard for quantitative assays.
  • Supplied in small research quantities appropriate for analytical applications.
  • Stable as a powder with recommended cold storage to preserve isotopic integrity.