Learn More
Medchemexpress LLC 2-{4-[(1S)-1-[(4,5-dichloro-1,6-dimethyl-1H-indol-2-yl)formamido]-2-hydroxyethyl]benzenesulfonyl}acetic acid | 2244452-09-1 | 99.6% | 499.36 g/mol | C21H20Cl2N2O6S | 100MG
SDP
Catalog No. 5000236985
Encompass_Preferred
BI-4924 is a selective, small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) used as a research tool to probe serine biosynthesis and metabolic dependencies in cells. It has low-nanomolar potency and favorable metabolic stability, and is supplied at high reported purity for biochemical and cellular studies.
  • Selective PHGDH inhibition with reported IC50 ≈ 3 nM.
  • High plasma protein binding and lipophilic profile.
  • High reported purity (99.6%).
  • Available as solid and as DMSO solution formats.
  • Molecular formula C21H20Cl2N2O6S; molecular weight 499.36 g/mol.
  • Supplied in multiple pack sizes, including 100 MG.
Catalog No. 50-002-36985 Supplier Medchemexpress LLC Supplier No. HY126254100MG
BI-4924 is a selective, small-molecule inhibitor of phosphoglycerate dehydrogenase (PHGDH) used as a research tool to probe serine biosynthesis and metabolic dependencies in cells. It has low-nanomolar potency and favorable metabolic stability, and is supplied at high reported purity for biochemical and cellular studies.
  • Selective PHGDH inhibition with reported IC50 ≈ 3 nM.
  • High plasma protein binding and lipophilic profile.
  • High reported purity (99.6%).
  • Available as solid and as DMSO solution formats.
  • Molecular formula C21H20Cl2N2O6S; molecular weight 499.36 g/mol.
  • Supplied in multiple pack sizes, including 100 MG.