Learn More
Medchemexpress LLC 2-[[4-(2,2-difluoropropoxy)pyrimidin-5-yl]methylamino]-4-[[(1R,4S)-4-hydroxy-3,3-dimethy... | 1799574-70-1 | 99.7% | C21H27F2N7O2 | 10MG
SDP
Catalog No. 5000260234
Encompass_Preferred
CC-90005 is a research-grade small-molecule inhibitor of protein kinase C-θ (PKC-θ). It is potent, selective, and orally active, commonly used in cellular and in vivo studies to probe PKC-θ-mediated signaling. Supplied as a white to off-white solid (typical pack: 10 mg) with formula C21H27F2N7O2 and molecular weight 447.48 g/mol. For research use only; not for human or clinical use.
  • Potent and selective PKC-θ inhibition (IC50 ~8 nM).
  • High purity suitable for analytical and biological studies.
  • Characterized by LC-MS and supplied with a certificate of analysis.
  • Suitable for cellular and in vivo pharmacology studies.
  • Stable solid form, easy to handle and store.
Catalog No. 50-002-60234 Supplier Medchemexpress LLC Supplier No. HY13230410MG
CC-90005 is a research-grade small-molecule inhibitor of protein kinase C-θ (PKC-θ). It is potent, selective, and orally active, commonly used in cellular and in vivo studies to probe PKC-θ-mediated signaling. Supplied as a white to off-white solid (typical pack: 10 mg) with formula C21H27F2N7O2 and molecular weight 447.48 g/mol. For research use only; not for human or clinical use.
  • Potent and selective PKC-θ inhibition (IC50 ~8 nM).
  • High purity suitable for analytical and biological studies.
  • Characterized by LC-MS and supplied with a certificate of analysis.
  • Suitable for cellular and in vivo pharmacology studies.
  • Stable solid form, easy to handle and store.