Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 2-[4-[2-(Fmoc-amino)ethyl]phenoxy]-2-methylpropanoic acid | 50 g | CAS 195131-58-9 | MDL MFCD20272674

Catalog No. 502352816
Encompass

2-[4-[2-(Fmoc-amino)ethyl]phenoxy]-2-methylpropanoic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 195131-58-9
- MDL: MFCD20272674
- InChIKey: GUNTZELRMUVRBA-UHFFFAOYSA-N
- Molecular Weight: 445.515
- Molecular Formula: C27H27NO5
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-(4-(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)ethyl)phenoxy)-2-methylpropanoic acid
- SMILES: CC(OC1=CC=C(C=C1)CCNC(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)(C(O)=O)C

Catalog No. 50-235-2816 Supplier STA Pharmaceutical Supplier No. STAAA1026750GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-[4-[2-(Fmoc-amino)ethyl]phenoxy]-2-methylpropanoic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 195131-58-9
- MDL: MFCD20272674
- InChIKey: GUNTZELRMUVRBA-UHFFFAOYSA-N
- Molecular Weight: 445.515
- Molecular Formula: C27H27NO5
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-(4-(2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)ethyl)phenoxy)-2-methylpropanoic acid
- SMILES: CC(OC1=CC=C(C=C1)CCNC(OCC2C3=CC=CC=C3C4=C2C=CC=C4)=O)(C(O)=O)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.