Learn More
Medchemexpress LLC 2-[[4-[(2-methoxybenzoyl)amino]benzoyl]amino]benzoic acid | 693241-54-2 | MFCD03624354 | 98.3% | 390.39 g/mol | C22H18N2O5 | 25MG
SDP
Catalog No. 5000236164
Encompass_Preferred
EG1 is a small-molecule inhibitor of the Pax2 transcription factor that binds the paired domain (Kd 1.35-1.5 μM) and disrupts Pax2-DNA interactions; provided as a solid research reagent for in vitro studies.
Catalog No. 50-002-36164 Supplier Medchemexpress LLC Supplier No. HY12483825MG
EG1 is a small-molecule inhibitor of the Pax2 transcription factor that binds the paired domain (Kd 1.35-1.5 μM) and disrupts Pax2-DNA interactions; provided as a solid research reagent for in vitro studies.