Learn More
Medchemexpress LLC 2-[[4-[(2-methoxybenzoyl)amino]benzoyl]amino]benzoic acid | 693241-54-2 | MFCD03624354 | 98.3% | 390.4 g·mol⁻1 | C22H18N2O5 | 50MG
SDP
Catalog No. 5000237072
Encompass_Preferred
EG1 is a small-molecule inhibitor of the Pax2 transcription factor that binds the Pax2 paired domain (Kd ≈ 1.35-1.5 μM) and inhibits Pax2-DNA interactions. It has been used to inhibit embryonic kidney development and to reduce viability of Pax2-positive renal cancer cell lines. Supplied as a purified research reagent with recommended storage conditions for powder and solutions.
  • Inhibits Pax2 DNA binding and transcriptional activity.
  • Demonstrated activity in developmental and renal cancer cell models.
  • High purity (98.3%) suitable for research applications.
  • Available in multiple pack sizes and a DMSO solution format.
  • Stable under recommended storage conditions for powder and in solvent.
Catalog No. 50-002-37072 Supplier Medchemexpress LLC Supplier No. HY12483850MG
EG1 is a small-molecule inhibitor of the Pax2 transcription factor that binds the Pax2 paired domain (Kd ≈ 1.35-1.5 μM) and inhibits Pax2-DNA interactions. It has been used to inhibit embryonic kidney development and to reduce viability of Pax2-positive renal cancer cell lines. Supplied as a purified research reagent with recommended storage conditions for powder and solutions.
  • Inhibits Pax2 DNA binding and transcriptional activity.
  • Demonstrated activity in developmental and renal cancer cell models.
  • High purity (98.3%) suitable for research applications.
  • Available in multiple pack sizes and a DMSO solution format.
  • Stable under recommended storage conditions for powder and in solvent.