Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-(2-(trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid | 1152782-19-8 | MFCD12912393 | 99.7% | 392.37 g/mol | C20H19F3N2O3 | 25 MG
SDP

Catalog No. 5000240256
Encompass_Preferred

A 1120 is a nonretinoid, high-affinity antagonist of retinol-binding protein 4 (RBP4). It binds RBP4 with a reported Ki of 8.3 nM and disrupts the interaction between RBP4 and transthyretin. The compound is provided as a solid research reagent with documented purity, solubility, and storage recommendations.

  • High-affinity RBP4 antagonist with Ki 8.3 nM.
  • Purity reported as 99.73%.
  • Molecular formula C20H19F3N2O3; molecular weight 392.37 g/mol.
  • Supplied as a solid suitable for research use.
  • Soluble in DMSO (100 mg/mL); in vivo vehicle example 10% DMSO, 90% corn oil.
  • Storage guidance provided for powder and solutions to maintain stability.

Catalog No. 50-002-40256 Supplier Medchemexpress LLC Supplier No. HY10763325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 1120 is a nonretinoid, high-affinity antagonist of retinol-binding protein 4 (RBP4). It binds RBP4 with a reported Ki of 8.3 nM and disrupts the interaction between RBP4 and transthyretin. The compound is provided as a solid research reagent with documented purity, solubility, and storage recommendations.

  • High-affinity RBP4 antagonist with Ki 8.3 nM.
  • Purity reported as 99.73%.
  • Molecular formula C20H19F3N2O3; molecular weight 392.37 g/mol.
  • Supplied as a solid suitable for research use.
  • Soluble in DMSO (100 mg/mL); in vivo vehicle example 10% DMSO, 90% corn oil.
  • Storage guidance provided for powder and solutions to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.